Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GPRASP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001099410.1 | NP_001092880.1 | 4 | 3937 | ||
NM_001099411.1 | NP_001092881.1 | 3 | 3869 | |||
NM_001184727.1 | NP_001171656.1 | 6 | 4040 | |||
NM_014710.4 | NP_055525.3 | 5 | 4028 | |||
XM_017029981.1 | XP_016885470.1 | 4 | 3917 | |||
XM_017029982.1 | XP_016885471.1 | 4 | 3952 | |||
GenBank mRNA | AB007903.2 | - | 1 | 3393 | ||
BC114552.1 | - | 3 | 3673 | 40037190 |