Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LAPTM4A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014713.4 | NP_055528.1 | 4 | 893 | ||
GenBank mRNA | AK129875.1 | - | 4 | 543 | ||
AK304433.1 | - | 4 | 461 | |||
AY359028.1 | - | 4 | 509 | |||
BC000421.2 | - | 4 | 470 | 2964450 | ||
BC003158.1 | - | 4 | 533 | 2958870 | ||
BC093075.1 | - | 4 | 493 | 30402963 | ||
CR456758.1 | - | 4 | 385 | |||
D14696.1 | - | 4 | 531 |