Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IFT140 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014714.3 | NP_055529.2 | 8 | 1226 | ||
XM_005255725.4 | XP_005255782.1 | 7 | 928 | |||
XM_005255726.3 | XP_005255783.1 | 7 | 928 | |||
XM_006720990.3 | XP_006721053.1 | 7 | 1144 | |||
XM_006720991.3 | XP_006721054.1 | 7 | 1334 | |||
XM_011522766.2 | XP_011521068.1 | 7 | 928 | |||
XM_011522769.2 | XP_011521071.1 | 7 | 928 | |||
XM_011522771.2 | XP_011521073.1 | 7 | 928 | |||
XM_011522772.2 | XP_011521074.1 | 7 | 928 | |||
XM_017023910.1 | XP_016879399.1 | 7 | 917 | |||
XM_017023911.1 | XP_016879400.1 | 6 | 895 | |||
GenBank mRNA | AB011162.1 | - | 7 | 923 | ||
AK001797.1 | - | 1271 | ||||
AK299035.1 | - | 7 | 925 | |||
BC035577.1 | - | 6 | 911 | 5552740 |