Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ARHGAP32 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001142685.1 | NP_001136157.1 | 22 | 4314 | ||
NM_014715.3 | NP_055530.2 | 13 | 3646 | |||
XM_005271736.4 | XP_005271793.2 | 23 | 5344 | |||
XM_011543072.2 | XP_011541374.1 | 23 | 4546 | |||
XM_011543073.2 | XP_011541375.2 | 23 | 4282 | |||
XM_011543075.2 | XP_011541377.1 | 14 | 3492 | |||
XM_017018595.1 | XP_016874084.1 | 23 | 4263 | |||
XM_017018596.1 | XP_016874085.1 | 21 | 4540 | |||
GenBank mRNA | AB018255.2 | - | 13 | 3567 | ||
AB088416.1 | - | 13 | 3567 | |||
AK025237.1 | - | 2 | 812 | |||
AL833062.2 | - | 19 | 6102 | |||
AY194287.1 | - | 13 | 3570 | |||
BC104898.1 | - | 12 | 3301 | 8143901 | ||
BC113429.1 | - | 12 | 3301 | 8322481 | ||
EF127492.1 | - | 22 | 4314 |