Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ACAP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014716.3 | NP_055531.1 | 10 | 1025 | ||
GenBank mRNA | BC018543.1 | - | 10 | 1000 | 4342840 | |
BT009788.1 | - | 10 | 819 | |||
D30758.1 | - | 10 | 1007 |