Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZFYVE16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001105251.2 | NP_001098721.1 | 9 | 3503 | ||
NM_001284236.1 | NP_001271165.1 | 9 | 3429 | |||
NM_014733.4 | NP_055548.3 | 8 | 3375 | |||
XM_005248632.4 | XP_005248689.1 | 8 | 3501 | |||
XM_011543753.2 | XP_011542055.1 | 9 | 3400 | |||
XM_017010091.1 | XP_016865580.1 | 10 | 3561 | |||
XM_017010092.1 | XP_016865581.1 | 9 | 3547 | |||
XM_017010093.1 | XP_016865582.1 | 8 | 1881 | |||
XM_017010094.1 | XP_016865583.1 | 7 | 1826 | |||
XM_017010095.1 | XP_016865584.1 | 10 | 3455 | |||
XM_017010096.1 | XP_016865585.1 | 5 | 903 | |||
GenBank mRNA | AB002303.1 | - | 9 | 3443 | ||
AF434817.1 | - | 7 | 3195 | |||
AK127003.1 | - | 4 | 593 | |||
AL833087.2 | - | 6 | 2422 | |||
BX537424.1 | - | 8 | 3327 | |||
CR933621.1 | - | 9 | 3378 |