Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZFYVE16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001105251.2 | NP_001098721.1 | 18, 19 | 4763 | ||
NM_001284236.1 | NP_001271165.1 | 18, 19 | 4689 | |||
NM_014733.4 | NP_055548.3 | 17, 18 | 4635 | |||
XM_005248632.4 | XP_005248689.1 | 17, 18 | 4761 | |||
XM_011543753.2 | XP_011542055.1 | 18, 19 | 4660 | |||
XM_017010091.1 | XP_016865580.1 | 19, 20 | 4821 | |||
XM_017010092.1 | XP_016865581.1 | 18, 19 | 4807 | |||
XM_017010093.1 | XP_016865582.1 | 17, 18 | 3141 | |||
XM_017010094.1 | XP_016865583.1 | 16, 17 | 3086 | |||
XM_017010095.1 | XP_016865584.1 | 19, 20 | 4715 | |||
XM_017010096.1 | XP_016865585.1 | 14, 15 | 2163 | |||
GenBank mRNA | AB002303.1 | - | 18, 19 | 4703 | ||
AF434817.1 | - | 16, 17 | 4455 | |||
AK127003.1 | - | 13, 14 | 1852 | |||
BX537424.1 | - | 17, 18 | 4587 | |||
BX649172.1 | - | 6, 7 | 3237 | |||
CR933621.1 | - | 18, 19 | 4637 |