Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RASSF2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014737.2 | NP_055552.1 | 11 | 1093 | ||
NM_170774.1 | NP_739580.1 | 10 | 951 | |||
XM_005260895.3 | XP_005260952.1 | 11 | 1159 | |||
XM_011529410.1 | XP_011527712.1 | 11 | 1093 | |||
XM_011529411.1 | XP_011527713.1 | 11 | 1165 | |||
XM_011529412.2 | XP_011527714.1 | 10 | 1014 | |||
XM_017028149.1 | XP_016883638.1 | 10 | 988 | |||
XM_017028150.1 | XP_016883639.1 | 12 | 1296 | |||
XM_017028151.1 | XP_016883640.1 | 12 | 1263 | |||
XM_017028152.1 | XP_016883641.1 | 12 | 1340 | |||
XM_017028153.1 | XP_016883642.1 | 10 | 1041 | |||
GenBank mRNA | AK222650.1 | - | 11 | 1056 | ||
AK291458.1 | - | 11 | 1048 | |||
AY154470.1 | - | 11 | 1093 | |||
AY154471.1 | - | 8 | 867 | |||
AY154472.1 | - | 10 | 951 | |||
BC110385.1 | - | 8 | 867 | |||
BC117118.1 | - | 9 | 930 | 40125669 | ||
BC117120.1 | - | 9 | 930 | 40125671 | ||
CR627436.1 | - | 10 | 4300 | |||
D79990.1 | - | 11 | 1093 |