Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EIF4A3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014740.3 | NP_055555.1 | 5 | 684 | ||
XM_011525522.1 | XP_011523824.1 | 4 | 577 | |||
GenBank mRNA | AK290608.1 | - | 5 | 665 | ||
BC003662.2 | - | 5 | 594 | 2988931 | ||
BC004386.1 | - | 5 | 615 | 3619386 | ||
BC011151.1 | - | 5 | 619 | 3452439 | ||
CR456750.1 | - | 5 | 462 | |||
D21853.1 | - | 5 | 676 | |||
X79538.1 | - | 5 | 535 |