Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TM9SF4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014742.3 | NP_055557.2 | 8 | 1064 | ||
XM_005260622.4 | XP_005260679.1 | 8 | 1116 | |||
XM_017028154.1 | XP_016883643.1 | 8 | 845 | |||
XM_017028155.1 | XP_016883644.1 | 8 | 1116 | |||
XM_017028156.1 | XP_016883645.1 | 8 | 812 | |||
XM_017028157.1 | XP_016883646.1 | 8 | 1069 | |||
XM_017028158.1 | XP_016883647.1 | 8 | 807 | |||
XM_017028159.1 | XP_016883648.1 | 8 | 869 | |||
XM_017028160.1 | XP_016883649.1 | 8 | 781 | |||
XM_017028161.1 | XP_016883650.1 | 8 | 817 | |||
GenBank mRNA | AK126534.1 | - | 6 | 514 | ||
AK294980.1 | - | 8 | 812 | |||
AK296500.1 | - | 8 | 721 | |||
AK302439.1 | - | 9 | 919 | |||
AK316293.1 | - | 7 | 768 | |||
BC021107.1 | - | 8 | 840 | 4896167 | ||
BC022850.2 | - | 7 | 822 | |||
D87444.1 | - | 8 | 1118 |