Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CDMO-CRO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other TBC1D5 SIRNA Assays ›
Gene Symbol:
TBC1D5
Gene Name:
TBC1 domain family member 5
Gene Aliases:
-
Chromosome Location:
Chr.3: 17157162 - 17742739 on Build GRCh38
UniGene ID:
Hs.475629
Species:
Human
siRNA ID
HSS114721
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: TBC1D5
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001134381.1
NP_001127853.1
8
657
NM_014744.2
NP_055559.1
7
2058
XM_005265611.1
XP_005265668.1
7
628
XM_005265612.1
XP_005265669.1
6
563
XM_005265614.1
XP_005265671.1
7
628
XM_005265615.1
XP_005265672.1
6
558
XM_005265616.4
XP_005265673.1
5
487
XM_006713430.1
XP_006713493.1
7
678
XM_011534281.1
XP_011532583.1
9
939
XM_011534283.2
XP_011532585.1
8
717
XM_011534284.2
XP_011532586.1
8
762
XM_011534286.1
XP_011532588.1
7
694
XM_011534287.1
XP_011532589.1
7
676
XM_017007552.1
XP_016863041.1
10
1040
XM_017007553.1
XP_016863042.1
10
1070
XM_017007554.1
XP_016863043.1
15
1683
XM_017007555.1
XP_016863044.1
9
922
XM_017007556.1
XP_016863045.1
8
902
XM_017007557.1
XP_016863046.1
6
522
XM_017007558.1
XP_016863047.1
6
552
XM_017007559.1
XP_016863048.1
8
902
XM_017007560.1
XP_016863049.1
8
732
XM_017007561.1
XP_016863050.1
6
563
XM_017007562.1
XP_016863051.1
7
676
XM_017007563.1
XP_016863052.1
5
482
XM_017007564.1
XP_016863053.1
6
558
XM_017007565.1
XP_016863054.1
7
856
XM_017007566.1
XP_016863055.1
7
645
XM_017007567.1
XP_016863056.1
5
486
XM_017007568.1
XP_016863057.1
6
522
GenBank mRNA
AB449886.1
-
5
393
AK309374.1
-
3
318
AK310539.1
-
8
657
BC013145.1
-
6
570
4510019
D86965.1
-
7
2057
Back To Top
More Information
Panther Classification:
Molecular Function -
GTPase-activating protein
Gene Ontology Categories:
Function(s)
Process(es)
positive regulation of receptor internalization
intracellular protein transport
positive regulation of autophagy
regulation of vesicle fusion
retrograde transport, endosome to Golgi
response to starvation
activation of GTPase activity
mitophagy in response to mitochondrial depolarization
regulation of cilium assembly
GTPase activator activity
protein binding
Rab GTPase binding
AP-2 adaptor complex binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-5c9c5c4749-vpmkj:80/100.66.78.144:80.
git-commit: d22414ba47b22cc02ffc24003f0e179da61511a6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.21.0-Offline