Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IST1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014761.3 | NP_055576.2 | 8, 9 | 959 | ||
GenBank mRNA | BC000430.2 | - | 8, 9 | 909 | 2819828 | |
BC004359.1 | - | 8, 9 | 909 | 2819828 |