Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EFCAB14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014774.2 | NP_055589.1 | 2 | 1285 | ||
GenBank mRNA | AB007963.1 | - | 2 | 1285 | ||
BC002525.2 | - | 2 | 430 | 3139273 | ||
BC018842.2 | - | 2 | 430 |