Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other MELK SIRNA Assays ›
Gene Symbol:
MELK
Gene Name:
maternal embryonic leucine zipper kinase
Gene Aliases:
HPK38
Chromosome Location:
Chr.9: 36572862 - 36677683 on Build GRCh38
UniGene ID:
Hs.184339
Species:
Human
siRNA ID
HSS114849
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: MELK
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001256685.1
NP_001243614.1
10
935
NM_001256687.1
NP_001243616.1
9
791
NM_001256688.1
NP_001243617.1
8
722
NM_001256689.1
NP_001243618.1
9
849
NM_001256690.1
NP_001243619.1
8
732
NM_001256691.1
NP_001243620.1
8
705
NM_001256692.1
NP_001243621.1
7
609
NM_001256693.1
NP_001243622.1
8
749
NM_014791.3
NP_055606.1
10
935
NR_046337.1
10
972
XM_011518076.2
XP_011516378.1
10
1114
XM_011518077.1
XP_011516379.1
11
1010
XM_011518078.2
XP_011516380.1
11
1197
XM_011518079.1
XP_011516381.1
10
887
XM_011518080.1
XP_011516382.1
10
902
XM_011518081.2
XP_011516383.1
10
920
XM_011518082.2
XP_011516384.1
9
1028
XM_011518083.2
XP_011516385.1
10
1111
XM_011518084.2
XP_011516386.1
9
723
XM_011518085.1
XP_011516387.1
8
719
XM_011518086.2
XP_011516388.1
10
902
GenBank mRNA
AB183427.1
-
9
800
AB183428.1
-
8
685
AK222811.1
-
10
934
AK293284.1
-
8
687
AK293423.1
-
8
670
AK293447.1
-
8
689
AK296820.1
-
10
971
AK299164.1
-
10
887
AK300761.1
-
9
756
AK300821.1
-
9
803
AK301131.1
-
8
681
AK302374.1
-
7
576
AK309470.1
-
9
808
BC014039.2
-
10
889
4547136
D79997.1
-
10
921
Back To Top
More Information
Panther Classification:
Molecular Function -
non-receptor serine/threonine protein kinase
Gene Ontology Categories:
Function(s)
Process(es)
G2/M transition of mitotic cell cycle
apoptotic process
cell proliferation
intrinsic apoptotic signaling pathway in response to oxidative stress
peptidyl-tyrosine phosphorylation
hemopoiesis
intracellular signal transduction
positive regulation of apoptotic process
protein autophosphorylation
neural precursor cell proliferation
protein serine/threonine kinase activity
non-membrane spanning protein tyrosine kinase activity
calcium ion binding
protein binding
ATP binding
lipid binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7487cff77c-x5whk:80/10.239.163.84:80.
git-commit: c531a7a49d92b854cb004d8acd820c41186dcba6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.2-2024.09.38-1.0