Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GAB2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012296.3 | NP_036428.1 | 6 | 1504 | ||
NM_080491.2 | NP_536739.1 | 6 | 1560 | |||
XM_006718753.2 | XP_006718816.1 | 7 | 1605 | |||
XM_011545408.2 | XP_011543710.1 | 5 | 1211 | |||
GenBank mRNA | AB011143.1 | - | 6 | 1504 | ||
AB018413.1 | - | 6 | 1458 | |||
BC131711.1 | - | 6 | 1462 | 40122341 | ||
BC152459.1 | - | 6 | 1504 | 8862595 |