Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KIAA0753 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014804.2 | NP_055619.2 | 10 | 2157 | ||
XM_006721612.2 | XP_006721675.1 | 8 | 1600 | |||
XM_011524090.2 | XP_011522392.1 | 10 | 2157 | |||
XM_011524091.2 | XP_011522393.1 | 10 | 3542 | |||
XM_011524095.2 | XP_011522397.1 | 10 | 1889 | |||
XM_011524096.1 | XP_011522398.1 | 10 | 2157 | |||
XM_011524098.2 | XP_011522400.1 | 8 | 1788 | |||
XM_011524099.2 | XP_011522401.1 | 8 | 1278 | |||
XM_011524100.2 | XP_011522402.1 | 7 | 1060 | |||
XM_017025455.1 | XP_016880944.1 | 10 | 2100 | |||
XM_017025456.1 | XP_016880945.1 | 8 | 1918 | |||
XM_017025457.1 | XP_016880946.1 | 7 | 1688 | |||
GenBank mRNA | AB018296.1 | - | 10 | 1902 | ||
AK292876.1 | - | 10 | 1919 | |||
AK296971.1 | - | 9 | 1799 | |||
AK300547.1 | - | 3 | 305 | |||
AL080108.1 | - | 4 | 590 | |||
BC017566.1 | - | 10 | 1916 | |||
BC113016.1 | - | 9 | 1872 | 40036441 | ||
BC113017.1 | - | 8 | 1765 | 40036446 |