Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LRIG2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001312686.1 | NP_001299615.1 | 18, 19 | 3313 | ||
NM_014813.2 | NP_055628.1 | 17, 18 | 3201 | |||
XM_005271369.2 | XP_005271426.1 | 16, 17 | 2880 | |||
XM_017002952.1 | XP_016858441.1 | 17, 18 | 2852 | |||
GenBank mRNA | AB018349.1 | - | 17, 18 | 3162 | ||
AK074825.1 | - | 5, 6 | 1237 | |||
AK123606.1 | - | 7, 8 | 2236 | |||
AK291031.1 | - | 17, 18 | 3193 | |||
BC117368.1 | - | 17, 18 | 3025 | 40125919 | ||
BC117370.1 | - | 17, 18 | 3025 | 40125921 | ||
CR936669.1 | - | 18, 19 | 3304 |