Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LRIG2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001312686.1 | NP_001299615.1 | 1, 2 | 477 | ||
NM_014813.2 | NP_055628.1 | 1, 2 | 477 | |||
XM_005271369.2 | XP_005271426.1 | 1, 2 | 477 | |||
GenBank mRNA | AB018349.1 | - | 1, 2 | 438 | ||
AK291031.1 | - | 1, 2 | 469 | |||
AL161998.1 | - | 1, 2 | 436 | |||
BC117368.1 | - | 1, 2 | 301 | 40125919 | ||
BC117370.1 | - | 1, 2 | 301 | 40125921 | ||
DA506094.1 | - | 1, 2 | 477 |