Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MAGI2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001301128.1 | NP_001288057.1 | 4 | 832 | ||
NM_012301.3 | NP_036433.2 | 4 | 832 | |||
XM_011516718.1 | XP_011515020.1 | 4 | 1063 | |||
XM_011516719.2 | XP_011515021.1 | 3 | 4351 | |||
XM_011516720.2 | XP_011515022.1 | 3 | 9341 | |||
XM_017012840.1 | XP_016868329.1 | 4 | 1063 | |||
XM_017012841.1 | XP_016868330.1 | 4 | 1063 | |||
XM_017012842.1 | XP_016868331.1 | 4 | 1063 | |||
XM_017012843.1 | XP_016868332.1 | 4 | 1063 | |||
XM_017012844.1 | XP_016868333.1 | 4 | 1063 | |||
XM_017012845.1 | XP_016868334.1 | 4 | 1063 | |||
XM_017012846.1 | XP_016868335.1 | 4 | 1063 | |||
XM_017012847.1 | XP_016868336.1 | 4 | 7300 | |||
XM_017012848.1 | XP_016868337.1 | 3 | 4348 | |||
XM_017012849.1 | XP_016868338.1 | 3 | 4348 | |||
XM_017012850.1 | XP_016868339.1 | 4 | 1063 | |||
XM_017012851.1 | XP_016868340.1 | 4 | 1063 | |||
XM_017012852.1 | XP_016868341.1 | 4 | 1063 | |||
GenBank mRNA | AB014605.2 | - | 4 | 813 | ||
AF038563.1 | - | 4 | 832 | |||
AK297353.1 | - | 4 | 817 | |||
AK297580.1 | - | 3 | 391 | |||
AK298988.1 | - | 4 | 707 | |||
BC150277.1 | - | 4 | 813 | 9007255 |