Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DDX46 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001300860.1 | NP_001287789.1 | 11 | 1579 | ||
NM_014829.3 | NP_055644.2 | 11 | 1579 | |||
NR_125341.1 | 10 | 1435 | ||||
XM_011543758.1 | XP_011542060.1 | 11 | 1542 | |||
GenBank mRNA | AB018344.1 | - | 11 | 1518 | ||
AK126011.1 | - | 10 | 1390 | |||
AK291788.1 | - | 11 | 1532 | |||
BC012304.2 | - | 11 | 1509 | 3873814 |