Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DDX46 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001300860.1 | NP_001287789.1 | 19 | 2705 | ||
NM_014829.3 | NP_055644.2 | 19 | 2705 | |||
NR_125341.1 | 19 | 2678 | ||||
GenBank mRNA | AB018344.1 | - | 19 | 2644 | ||
AF106680.1 | - | 19 | 2645 | |||
AK126011.1 | - | 19 | 2633 | |||
AK291788.1 | - | 19 | 2658 | |||
BC012304.2 | - | 19 | 2635 | 3873814 | ||
BX647426.1 | - | 7 | 1364 |