Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RBM19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001146698.1 | NP_001140170.1 | 11 | 1494 | ||
NM_001146699.1 | NP_001140171.1 | 11 | 1494 | |||
NM_016196.3 | NP_057280.2 | 11 | 1494 | |||
XM_017020280.1 | XP_016875769.1 | 10 | 1422 | |||
XM_017020281.1 | XP_016875770.1 | 10 | 1422 | |||
GenBank mRNA | AB014582.1 | - | 11 | 1429 | ||
AK291444.1 | - | 11 | 1445 | |||
AK314606.1 | - | 11 | 1494 | |||
AL117547.1 | - | 11 | 1435 | |||
BC004289.1 | - | 11 | 1433 | 3619715 | ||
BC006137.1 | - | 11 | 1433 | 3677625 |