Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: G3BP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012297.4 | NP_036429.2 | 7 | 733 | ||
NM_203504.2 | NP_987100.1 | 7 | 868 | |||
NM_203505.2 | NP_987101.1 | 7 | 868 | |||
XM_005263382.2 | XP_005263439.1 | 7 | 842 | |||
XM_005263383.3 | XP_005263440.1 | 8 | 807 | |||
XM_011532441.2 | XP_011530743.1 | 7 | 728 | |||
XM_017008876.1 | XP_016864365.1 | 7 | 716 | |||
XM_017008877.1 | XP_016864366.1 | 7 | 895 | |||
XM_017008878.1 | XP_016864367.1 | 7 | 716 | |||
XM_017008879.1 | XP_016864368.1 | 7 | 742 | |||
GenBank mRNA | AB014560.1 | - | 7 | 688 | ||
AF051311.1 | - | 6 | 568 | |||
AF053535.1 | - | 7 | 763 | |||
AF070615.1 | - | 3 | 168 | |||
AF145284.1 | - | 7 | 763 | |||
AK291772.1 | - | 7 | 798 | |||
AK291786.1 | - | 7 | 703 | |||
BC011731.2 | - | 7 | 786 | 4337457 | ||
CB996973.1 | - | 7 | 719 |