Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ARHGAP44 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001321164.1 | NP_001308093.1 | 11, 12 | 1301 | ||
NM_001321166.1 | NP_001308095.1 | 11, 12 | 1301 | |||
NM_001321167.1 | NP_001308096.1 | 11, 12 | 1301 | |||
NM_001321168.1 | NP_001308097.1 | 10, 11 | 1183 | |||
NM_014859.5 | NP_055674.4 | 11, 12 | 1301 | |||
NR_135569.1 | 11, 12 | 1301 | ||||
XM_005256891.1 | XP_005256948.1 | 11, 12 | 1151 | |||
GenBank mRNA | AB014572.1 | - | 11, 12 | 1274 | ||
AK294538.1 | - | 11, 12 | 1046 | |||
BC070074.1 | - | 10, 11 | 961 | |||
BC094754.1 | - | 10, 11 | 961 | |||
BC117412.1 | - | 11, 12 | 1053 | 40125963 | ||
BC117416.1 | - | 11, 12 | 1053 | 40125967 | ||
BC143852.1 | - | 11, 12 | 1053 | |||
BC143853.1 | - | 11, 12 | 1053 | 9052369 | ||
BC143855.1 | - | 11, 12 | 1053 | |||
BC143856.1 | - | 11, 12 | 1053 |