Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RNF10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014868.4 | NP_055683.3 | 7 | 1519 | ||
XM_005254013.2 | XP_005254070.1 | 8 | 1596 | |||
XM_006719717.2 | XP_006719780.1 | 7 | 1539 | |||
XM_017020282.1 | XP_016875771.1 | 8 | 1596 | |||
XM_017020283.1 | XP_016875772.1 | 7 | 1539 | |||
GenBank mRNA | AB027196.1 | - | 7 | 1483 | ||
AK094011.1 | - | 8 | 1420 | |||
AK126274.1 | - | 8 | 1324 | |||
AK301608.1 | - | 8 | 1393 | |||
AK315558.1 | - | 7 | 1501 | |||
AL389976.1 | - | 6 | 776 | |||
BC016622.1 | - | 7 | 1483 | 3874214 | ||
BC031596.1 | - | 7 | 1483 | 5177665 | ||
BC101709.1 | - | 7 | 1109 | 8069215 | ||
BC101715.1 | - | 7 | 1109 | 8069221 | ||
BC143493.1 | - | 7 | 1108 | |||
BT006974.1 | - | 7 | 1036 | |||
D87451.2 | - | 7 | 1036 |