Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RNF10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014868.4 | NP_055683.3 | 16 | 2815 | ||
XM_005254013.2 | XP_005254070.1 | 17 | 2907 | |||
XM_006719717.2 | XP_006719780.1 | 16 | 2850 | |||
XM_017020282.1 | XP_016875771.1 | 17 | 2892 | |||
GenBank mRNA | AB027196.1 | - | 16 | 2779 | ||
AK094011.1 | - | 17 | 2716 | |||
AK097807.1 | - | 17 | 2741 | |||
AK126274.1 | - | 17 | 2620 | |||
AK301608.1 | - | 17 | 2689 | |||
AK315558.1 | - | 16 | 2797 | |||
AL389976.1 | - | 15 | 2087 | |||
BC016622.1 | - | 16 | 2779 | 3874214 | ||
BC031596.1 | - | 16 | 2779 | 5177665 | ||
BC101709.1 | - | 16 | 2405 | 8069215 | ||
BC101715.1 | - | 16 | 2405 | 8069221 | ||
BC143493.1 | - | 15 | 2303 | |||
BT006974.1 | - | 16 | 2332 | |||
D87451.2 | - | 16 | 2332 |