Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IQSEC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001134382.2 | NP_001127854.1 | 3 | 1614 | ||
NM_014869.6 | NP_055684.3 | 3 | 1800 | |||
XM_011534304.2 | XP_011532606.1 | 5 | 1851 | |||
XM_011534305.2 | XP_011532607.1 | 3 | 1600 | |||
XM_011534306.2 | XP_011532608.1 | 3 | 2006 | |||
XM_011534307.2 | XP_011532609.1 | 4 | 1952 | |||
XM_011534308.2 | XP_011532610.1 | 3 | 1795 | |||
XM_011534310.2 | XP_011532612.1 | 5 | 1851 | |||
XM_011534311.2 | XP_011532613.1 | 3 | 1963 | |||
XM_011534312.2 | XP_011532614.1 | 3 | 1591 | |||
XM_011534313.2 | XP_011532615.1 | 3 | 1810 | |||
XM_011534314.2 | XP_011532616.1 | 3 | 1546 | |||
XM_011534315.2 | XP_011532617.1 | 3 | 1549 | |||
XM_017007581.1 | XP_016863070.1 | 5 | 1851 | |||
XM_017007582.1 | XP_016863071.1 | 4 | 1806 | |||
GenBank mRNA | AB018306.1 | - | 2 | 1293 | ||
AK294525.1 | - | 3 | 1614 | |||
AY653734.1 | - | 3 | 1562 | |||
BC010267.1 | - | 3 | 1554 | 2900367 |