Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZBTB40 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001083621.1 | NP_001077090.1 | 6 | 1658 | ||
NM_014870.3 | NP_055685.3 | 5 | 1370 | |||
XM_011542499.2 | XP_011540801.1 | 5 | 1393 | |||
GenBank mRNA | AB007947.2 | - | 5 | 1364 | ||
AK091019.1 | - | 6 | 1658 | |||
AK095273.1 | - | 5 | 1730 | |||
DA248977.1 | - | 3 | 448 |