Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZBTB5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014872.2 | NP_055687.1 | 2 | 485 | ||
XM_005251634.1 | XP_005251691.1 | 2 | 1329 | |||
GenBank mRNA | AB002352.1 | - | 2 | 474 | ||
AK292404.1 | - | 2 | 530 | |||
BC010007.2 | - | 2 | 329 | 4300173 |