Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MFN2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001127660.1 | NP_001121132.1 | 15 | 2156 | ||
NM_014874.3 | NP_055689.1 | 16 | 2301 | |||
XM_005263543.3 | XP_005263600.1 | 17 | 2314 | |||
XM_005263545.3 | XP_005263602.1 | 16 | 2164 | |||
XM_005263547.3 | XP_005263604.1 | 16 | 2196 | |||
XM_005263548.3 | XP_005263605.1 | 17 | 2331 | |||
GenBank mRNA | AF036536.2 | - | 16 | 2170 | ||
AK001189.1 | - | 5 | 637 | |||
AK021947.1 | - | 10 | 1336 | |||
AK022453.1 | - | 6 | 810 | |||
AK289828.1 | - | 15 | 2101 | |||
AK295890.1 | - | 14 | 1628 | |||
AL137666.1 | - | 2 | 189 | |||
AY028429.1 | - | 15 | 1874 | |||
BC017061.1 | - | 16 | 2031 | 3901235 | ||
D86987.1 | - | 16 | 2170 |