Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ARHGAP25 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001007231.2 | NP_001007232.2 | 8 | 1320 | ||
NM_001166276.1 | NP_001159748.1 | 7 | 1260 | |||
NM_001166277.1 | NP_001159749.1 | 7 | 1200 | |||
NM_014882.2 | NP_055697.1 | 7 | 1257 | |||
XM_005264675.2 | XP_005264732.1 | 8 | 1024 | |||
XM_005264676.2 | XP_005264733.1 | 7 | 908 | |||
XM_011533206.2 | XP_011531508.1 | 8 | 1068 | |||
XM_011533207.2 | XP_011531509.1 | 8 | 1034 | |||
XM_011533208.2 | XP_011531510.1 | 9 | 1085 | |||
XM_011533209.2 | XP_011531511.1 | 5 | 625 | |||
XM_011533210.2 | XP_011531512.1 | 3 | 152 | |||
XM_017005423.1 | XP_016860912.1 | 8 | 928 | |||
XM_017005424.1 | XP_016860913.1 | 8 | 925 | |||
XM_017005425.1 | XP_016860914.1 | 7 | 811 | |||
XM_017005426.1 | XP_016860915.1 | 5 | 1262 | |||
XM_017005427.1 | XP_016860916.1 | 5 | 1259 | |||
GenBank mRNA | AK290396.1 | - | 8 | 998 | ||
AK297056.1 | - | 7 | 1078 | |||
AK297157.1 | - | 8 | 1060 | |||
AK298669.1 | - | 7 | 1087 | |||
AK311669.1 | - | 8 | 1259 | |||
BC039591.1 | - | 7 | 1121 | 5757311 | ||
D29642.1 | - | 7 | 1070 | |||
EU794656.1 | - | 7 | 1257 |