Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ARHGAP25 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001007231.2 | NP_001007232.2 | 10 | 1877 | ||
NM_001166276.1 | NP_001159748.1 | 9 | 1817 | |||
NM_001166277.1 | NP_001159749.1 | 9 | 1757 | |||
NM_014882.2 | NP_055697.1 | 9 | 1814 | |||
XM_005264675.2 | XP_005264732.1 | 10 | 1581 | |||
XM_005264676.2 | XP_005264733.1 | 9 | 1465 | |||
XM_011533206.2 | XP_011531508.1 | 10 | 1625 | |||
XM_011533207.2 | XP_011531509.1 | 10 | 1591 | |||
XM_011533208.2 | XP_011531510.1 | 11 | 1642 | |||
XM_011533209.2 | XP_011531511.1 | 7 | 1182 | |||
XM_011533210.2 | XP_011531512.1 | 5 | 709 | |||
XM_017005423.1 | XP_016860912.1 | 10 | 1485 | |||
XM_017005424.1 | XP_016860913.1 | 10 | 1482 | |||
XM_017005425.1 | XP_016860914.1 | 9 | 1368 | |||
XM_017005426.1 | XP_016860915.1 | 7 | 1819 | |||
XM_017005427.1 | XP_016860916.1 | 7 | 1816 | |||
GenBank mRNA | AK290396.1 | - | 10 | 1555 | ||
AK297056.1 | - | 9 | 1635 | |||
AK297157.1 | - | 10 | 1617 | |||
AK298669.1 | - | 9 | 1644 | |||
D29642.1 | - | 9 | 1627 | |||
EU794656.1 | - | 9 | 1814 |