Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AMMECR1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001025580.1 | NP_001020751.1 | 1 | 466 | ||
NM_001171689.1 | NP_001165160.1 | 3 | 653 | |||
NM_015365.2 | NP_056180.1 | 1 | 466 | |||
GenBank mRNA | AJ007014.1 | - | 1 | 553 | ||
AK091430.1 | - | 3 | 653 | |||
AK314596.1 | - | 1 | 397 | |||
BC060813.1 | - | 1 | 399 | 30344824 | ||
BX648797.1 | - | 1 | 428 | |||
DA203031.1 | - | 3 | 428 | |||
KU178496.1 | - | 1 | 385 | |||
KU178497.1 | - | 1 | 385 |