Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GOLGA5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005113.3 | NP_005104.3 | 9, 10 | 1973 | ||
XM_011537420.2 | XP_011535722.1 | 9, 10 | 1973 | |||
GenBank mRNA | AF085199.1 | - | 9, 10 | 1848 | ||
AJ132949.1 | - | 8, 9 | 1717 | |||
AK293077.1 | - | 9, 10 | 1876 | |||
AK303789.1 | - | 10, 11 | 1606 | |||
AY644768.1 | - | 8, 9 | 1753 | |||
BC023021.1 | - | 9, 10 | 1881 | 4999710 |