Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HS3ST4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006040.2 | NP_006031.2 | 2 | 1329 | ||
GenBank mRNA | AF105378.2 | - | 2 | 1326 | ||
AL834283.1 | - | 2 | 576 | |||
EF063601.1 | - | 2 | 400 | |||
EF063602.1 | - | 2 | 400 | |||
EF063603.1 | - | 2 | 400 | |||
EF063604.1 | - | 2 | 400 | |||
EF063605.1 | - | 2 | 400 | |||
EF063606.1 | - | 2 | 400 | |||
EF063607.1 | - | 2 | 400 | |||
EF063608.1 | - | 2 | 400 |