Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HS3ST2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006043.1 | NP_006034.1 | 1 | 468 | ||
XM_011546001.2 | XP_011544303.1 | 1 | 845 | |||
XM_011546002.2 | XP_011544304.1 | 1 | 844 | |||
GenBank mRNA | AF105374.1 | - | 1 | 468 | ||
AF105375.1 | - | 1 | 468 | |||
AY359095.2 | - | 1 | 831 | |||
BC093734.1 | - | 1 | 432 | 7939579 | ||
BC093736.1 | - | 1 | 431 | 7939581 |