Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: USP15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001252078.1 | NP_001239007.1 | 3 | 346 | ||
NM_001252079.1 | NP_001239008.1 | 3 | 346 | |||
NM_006313.2 | NP_006304.1 | 3 | 346 | |||
XM_006719718.3 | XP_006719781.1 | 3 | 344 | |||
XM_017020285.1 | XP_016875774.1 | 3 | 297 | |||
GenBank mRNA | AB011101.2 | - | 3 | 271 | ||
AF013990.1 | - | 3 | 217 | |||
AF106069.1 | - | 3 | 280 | |||
AF153604.1 | - | 3 | 328 | |||
AK023703.1 | - | 3 | 289 | |||
AK292337.1 | - | 3 | 306 | |||
AK295947.1 | - | 3 | 278 | |||
AK297759.1 | - | 3 | 282 | |||
BC020688.1 | - | 3 | 282 | |||
BC063454.1 | - | 3 | 286 | |||
BC110908.1 | - | 3 | 279 | |||
BC125123.1 | - | 3 | 280 | 40118994 | ||
DB451322.1 | - | 3 | 348 |