Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TNFSF15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005118.3 | NP_005109.2 | 1 | 155 | ||
GenBank mRNA | AF520785.1 | - | 1 | 161 | ||
AK291642.1 | - | 1 | 66 | |||
AK313721.1 | - | 1 | 130 | |||
BC069435.1 | - | 1 | 110 | 7262219 | ||
BC074940.2 | - | 1 | 79 | 30915494 | ||
BC074941.2 | - | 1 | 79 | 30915271 | ||
BC104462.1 | - | 1 | 154 | 40030800 | ||
BC104463.1 | - | 1 | 155 | 40030803 |