Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NR1H4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001206977.1 | NP_001193906.1 | 11 | 1805 | ||
NM_001206978.1 | NP_001193907.1 | 9 | 1485 | |||
NM_001206979.1 | NP_001193908.1 | 10 | 1638 | |||
NM_001206992.1 | NP_001193921.1 | 8 | 1208 | |||
NM_001206993.1 | NP_001193922.1 | 8 | 1220 | |||
NM_005123.3 | NP_005114.1 | 10 | 1626 | |||
NR_135146.1 | 10 | 1631 | ||||
XM_011539040.2 | XP_011537342.1 | 10 | 2556 | |||
XM_011539041.2 | XP_011537343.1 | 7 | 1465 | |||
GenBank mRNA | AB307699.1 | - | 8 | 1150 | ||
AF384555.1 | - | 8 | 1162 | |||
AF478445.1 | - | 8 | 1208 | |||
AF478446.1 | - | 8 | 1220 | |||
AK296612.1 | - | 8 | 1221 | |||
AK296647.1 | - | 8 | 1156 | |||
AK312959.1 | - | 10 | 1627 | |||
BC071778.1 | - | 8 | 1160 | |||
BC130573.1 | - | 10 | 1495 | 40146604 | ||
BC144183.1 | - | 9 | 1343 | |||
BC144184.1 | - | 10 | 1495 | 9052704 | ||
BX571749.1 | - | 8 | 1205 | |||
HQ709175.1 | - | 8 | 1162 | |||
U68233.1 | - | 10 | 1503 |