Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RBX1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014248.3 | NP_055063.1 | 2 | 168 | ||
GenBank mRNA | AF085906.1 | - | 2 | 97 | ||
AF140598.1 | - | 2 | 122 | |||
AF142059.1 | - | 2 | 116 | |||
AK315722.1 | - | 2 | 127 | |||
AY099360.1 | - | 2 | 79 | |||
BC001466.2 | - | 2 | 121 | 3138751 | ||
BC017370.1 | - | 2 | 138 | |||
CR456560.1 | - | 2 | 134 | |||
DA353856.1 | - | 2 | 168 |