Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC12A6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001042494.1 | NP_001035959.1 | 18 | 2573 | ||
NM_001042495.1 | NP_001035960.1 | 18 | 2248 | |||
NM_001042496.1 | NP_001035961.1 | 18 | 2639 | |||
NM_001042497.1 | NP_001035962.1 | 16 | 2342 | |||
NM_005135.2 | NP_005126.1 | 17 | 2212 | |||
NM_133647.1 | NP_598408.1 | 17 | 2387 | |||
XM_006720793.3 | XP_006720856.1 | 16 | 3122 | |||
XM_011522269.2 | XP_011520571.1 | 16 | 3149 | |||
GenBank mRNA | AF105366.1 | - | 17 | 2387 | ||
AF108831.1 | - | 17 | 2121 | |||
AF116242.1 | - | 17 | 2223 | |||
AF477977.1 | - | 18 | 2580 | |||
AF531258.1 | - | 16 | 2342 | |||
AF531259.1 | - | 18 | 2580 | |||
AF531260.1 | - | 18 | 2218 | |||
AK128133.1 | - | 16 | 1895 | |||
AK292550.1 | - | 18 | 2326 | |||
AK315283.1 | - | 17 | 2184 | |||
AL117500.1 | - | 14 | 1815 | |||
BC051744.1 | - | 18 | 2576 | 4829214 | ||
BC070107.1 | - | 18 | 2243 | 4826538 | ||
BC126241.1 | - | 17 | 2120 | 8991957 | ||
BC126243.1 | - | 17 | 2120 | 8991959 | ||
BX648195.1 | - | 17 | 3329 | |||
DQ138323.1 | - | 18 | 2638 |