Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DGCR2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001173533.1 | NP_001167004.1 | 5 | 900 | ||
NM_001173534.1 | NP_001167005.1 | 5 | 891 | |||
NM_001184781.1 | NP_001171710.1 | 6 | 1014 | |||
NM_005137.2 | NP_005128.1 | 6 | 1023 | |||
NR_033674.1 | 5 | 908 | ||||
GenBank mRNA | AK291670.1 | - | 6 | 981 | ||
AK295697.1 | - | 5 | 866 | |||
AK296337.1 | - | 5 | 849 | |||
AK303130.1 | - | 6 | 933 | |||
AK304382.1 | - | 5 | 858 | |||
BC020339.1 | - | 6 | 970 | |||
BC032430.1 | - | 6 | 959 | 5243412 | ||
BC035425.1 | - | 6 | 970 | |||
BC040500.1 | - | 6 | 1018 | 5272271 | ||
CR456433.1 | - | 6 | 795 | |||
CR936871.1 | - | 7 | 1358 | |||
D79985.1 | - | 6 | 960 | |||
FJ032369.1 | - | 6 | 775 | |||
L77560.1 | - | 6 | 924 | |||
X83545.1 | - | 6 | 924 | |||
X84076.1 | - | 6 | 921 |