Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AKT3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001206729.1 | NP_001193658.1 | 3 | 286 | ||
NM_005465.4 | NP_005456.1 | 2 | 245 | |||
NM_181690.2 | NP_859029.1 | 2 | 245 | |||
XM_005272994.4 | XP_005273051.1 | 3 | 470 | |||
XM_005272995.2 | XP_005273052.1 | 3 | 276 | |||
XM_006711726.3 | XP_006711789.1 | 3 | 470 | |||
XM_011544012.2 | XP_011542314.1 | 3 | 470 | |||
XM_011544013.2 | XP_011542315.1 | 3 | 470 | |||
XM_016999985.1 | XP_016855474.1 | 3 | 304 | |||
GenBank mRNA | AF085234.1 | - | 2 | 180 | ||
AF124141.1 | - | 2 | 145 | |||
AF135794.1 | - | 2 | 133 | |||
AJ245709.1 | - | 2 | 143 | |||
AK308052.1 | - | 2 | 171 | |||
AL117525.1 | - | 3 | 286 | |||
AW960221.1 | - | 2 | 225 | |||
AY005799.1 | - | 2 | 206 | |||
BC121154.2 | - | 2 | 216 | 40121374 |