Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MED6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001284209.1 | NP_001271138.1 | 5 | 521 | ||
NM_001284210.1 | NP_001271139.1 | 5 | 500 | |||
NM_001284211.1 | NP_001271140.1 | 5 | 500 | |||
NM_005466.3 | NP_005457.2 | 5 | 500 | |||
GenBank mRNA | AF155104.1 | - | 4 | 422 | ||
AK222587.1 | - | 5 | 476 | |||
AK223346.1 | - | 5 | 465 | |||
AK290647.1 | - | 5 | 465 | |||
AK300417.1 | - | 4 | 772 | |||
AK300460.1 | - | 5 | 486 | |||
AK304361.1 | - | 5 | 465 | |||
BC004106.2 | - | 5 | 474 | 3936675 | ||
BM802752.1 | - | 452 | ||||
BT006831.1 | - | 5 | 447 | |||
CR541697.1 | - | 5 | 447 | |||
U78082.1 | - | 5 | 497 |