Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PDCD6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001267556.1 | NP_001254485.1 | 5 | 494 | ||
NM_001267557.1 | NP_001254486.1 | 3 | 296 | |||
NM_001267558.1 | NP_001254487.1 | 6 | 415 | |||
NM_013232.3 | NP_037364.1 | 5 | 500 | |||
GenBank mRNA | AF035606.1 | - | 5 | 526 | ||
AK001917.1 | - | 5 | 445 | |||
AK124302.1 | - | 1 | 1755 | |||
AK128087.1 | - | 3381 | ||||
AK223366.1 | - | 5 | 500 | |||
AK308872.1 | - | 4 | 398 | |||
AK315370.1 | - | 5 | 500 | |||
BC012384.1 | - | 5 | 468 | 3859672 | ||
BC106706.2 | - | 5 | 449 | 40003290 | ||
BC110291.1 | - | 5 | 458 | 5187550 | ||
BG390506.1 | - | 3 | 226 | |||
BI553617.1 | - | 5 | 496 | |||
BT020072.1 | - | 5 | 400 | |||
DB495449.1 | - | 6 | 400 | |||
U58773.1 | - | 5 | 431 |