Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MED16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005481.2 | NP_005472.2 | 11 | 1948 | ||
XM_017026120.1 | XP_016881609.1 | 10 | 1736 | |||
XM_017026121.1 | XP_016881610.1 | 10 | 1788 | |||
GenBank mRNA | AF106934.1 | - | 11 | 1935 | ||
AF121228.1 | - | 10 | 1797 | |||
AL137732.1 | - | 4 | 650 | |||
BC004554.1 | - | 11 | 1929 | 4053743 | ||
BC007853.2 | - | 7 | 1058 | |||
BC011841.1 | - | 11 | 1929 | 4053664 | ||
BC017282.2 | - | 11 | 1923 | 4876977 | ||
EU392449.1 | - | 9 | 1607 | |||
EU392451.1 | - | 9 | 1376 | |||
EU392452.1 | - | 7 | 1097 | |||
EU392453.1 | - | 7 | 1091 | |||
EU392454.1 | - | 7 | 968 | |||
EU392455.1 | - | 5 | 533 | |||
EU392456.1 | - | 5 | 578 | |||
EU392457.1 | - | 4 | 491 | |||
EU392458.1 | - | 4 | 407 | |||
EU392459.1 | - | 4 | 398 | |||
EU392460.1 | - | 3 | 287 | |||
EU392461.1 | - | 3 | 137 | |||
EU392462.1 | - | 2 | 256 | |||
EU392463.1 | - | 2 | 155 | |||
EU392484.1 | - | 6 | 723 | |||
EU392485.1 | - | 6 | 562 | |||
EU392486.1 | - | 5 | 421 | |||
EU392487.1 | - | 5 | 633 | |||
EU392488.1 | - | 5 | 366 | |||
EU392489.1 | - | 5 | 492 | |||
EU392490.1 | - | 4 | 291 | |||
EU392491.1 | - | 3 | 172 | |||
EU392492.1 | - | 4 | 255 | |||
EU392493.1 | - | 2 | 261 | |||
EU392494.1 | - | 3 | 339 | |||
EU392495.1 | - | 2 | 262 | |||
EU392496.1 | - | 4 | 205 |