Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MED16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005481.2 | NP_005472.2 | 2 | 207 | ||
XM_017026120.1 | XP_016881609.1 | 2 | 202 | |||
XM_017026121.1 | XP_016881610.1 | 2 | 217 | |||
GenBank mRNA | AF106934.1 | - | 2 | 194 | ||
AF121228.1 | - | 1 | 56 | |||
AU098434.1 | - | 2 | 207 | |||
BC004554.1 | - | 2 | 188 | 4053743 | ||
BC011841.1 | - | 2 | 188 | 4053664 | ||
BC017282.2 | - | 2 | 182 | 4876977 | ||
BI461485.1 | - | 2 | 209 | |||
EU392449.1 | - | 1 | 73 | |||
EU392451.1 | - | 1 | 73 | |||
EU392452.1 | - | 1 | 73 | |||
EU392453.1 | - | 1 | 73 | |||
EU392454.1 | - | 1 | 73 | |||
EU392456.1 | - | 1 | 73 | |||
EU392457.1 | - | 1 | 73 | |||
EU392459.1 | - | 1 | 73 | |||
EU392460.1 | - | 1 | 73 | |||
EU392464.1 | - | 1 | 73 | |||
EU392465.1 | - | 1 | 73 | |||
EU392466.1 | - | 1 | 73 | |||
EU392467.1 | - | 1 | 73 | |||
EU392468.1 | - | 1 | 73 | |||
EU392469.1 | - | 1 | 73 | |||
EU392470.1 | - | 1 | 73 | |||
EU392471.1 | - | 1 | 73 | |||
EU392472.1 | - | 1 | 73 | |||
EU392473.1 | - | 1 | 73 | |||
EU392475.1 | - | 1 | 73 | |||
EU392476.1 | - | 1 | 73 | |||
EU392477.1 | - | 1 | 73 | |||
EU392478.1 | - | 1 | 73 | |||
EU392480.1 | - | 1 | 73 | |||
EU392484.1 | - | 1 | 73 | |||
EU392486.1 | - | 1 | 73 | |||
EU392487.1 | - | 1 | 73 | |||
EU392494.1 | - | 1 | 73 |