Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CHAF1A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005483.2 | NP_005474.2 | 8 | 1682 | ||
XM_011527605.2 | XP_011525907.1 | 8 | 1749 | |||
XM_011527606.2 | XP_011525908.1 | 8 | 1749 | |||
XM_011527607.2 | XP_011525909.1 | 7 | 1692 | |||
GenBank mRNA | BC067093.1 | - | 8 | 1683 | 6339027 | |
U20979.1 | - | 8 | 1568 |