Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PARP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001042618.1 | NP_001036083.1 | 9 | 913 | ||
NM_005484.3 | NP_005475.2 | 9 | 952 | |||
XM_005267247.3 | XP_005267304.1 | 9 | 952 | |||
XM_017020912.1 | XP_016876401.1 | 9 | 1486 | |||
GenBank mRNA | AF085734.1 | - | 9 | 949 | ||
AJ236912.1 | - | 9 | 887 | |||
AK001980.1 | - | 9 | 929 | |||
AK290079.1 | - | 9 | 892 | |||
AK300971.1 | - | 7 | 670 |