Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SH2D3A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005490.2 | NP_005481.2 | 4 | 635 | ||
XM_005259467.1 | XP_005259524.1 | 4 | 585 | |||
XM_005259472.2 | XP_005259529.1 | 3 | 198 | |||
XM_006722602.1 | XP_006722665.1 | 4 | 585 | |||
XM_011527608.2 | XP_011525910.1 | 3 | 1002 | |||
XM_011527609.2 | XP_011525911.1 | 3 | 499 | |||
XM_017026123.1 | XP_016881612.1 | 4 | 585 | |||
XM_017026124.1 | XP_016881613.1 | 4 | 585 | |||
GenBank mRNA | AF124249.1 | - | 4 | 592 | ||
AK024368.1 | - | 3 | 1274 | |||
AK225386.1 | - | 4 | 558 | |||
AK290293.1 | - | 4 | 711 | |||
AK292781.1 | - | 4 | 558 | |||
AK299406.1 | - | 3 | 208 | |||
AK310659.1 | - | 4 | 558 | |||
AY358406.1 | - | 4 | 592 | |||
BC006281.2 | - | 4 | 572 | 3958342 |